HS4283P
Microtube Storage Box
to hold, 50 tube(s), white expanded polystyrene rack
All Photos(1)
About This Item
UNSPSC Code:
41122800
Recommended Products
material
white expanded polystyrene rack
feature
to hold 50 tube(s)
packaging
pack of 10 ea
manufacturer/tradename
Heathrow Scientific HS4283P
L × W × H
210 mm × 110 mm × 71 mm
General description
The Foam Tube Rack provides high storage with minimal footprint.
Features and Benefits
- Two-piece racks are constructed of expanded polystyrene (EPS) foam
- Lightweight microtube racks are ideal for long-term studies or tube storage
- Extra spacing between wells allows for easy gripping with fingertips
- Racks stack securely with nesting features on lid and base
- Store down to temperatures of -80°C. Must use tubes capable of being stored at the required temperature of the user
- Not recommended for autoclaving
Choose from one of the most recent versions:
Certificates of Analysis (COA)