Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

A1352

Sigma-Aldrich

AMMC iodide

≥99% purity (NMR), solid

Synonym(s):

3-[2-(N,N-diethyl-N-methylammonium)ethyl]-7-methoxy-4-methylcoumarin iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26INO3
Molecular Weight:
431.31
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

AMMC iodide, ≥99% (NMR)

Quality Level

Assay

≥99% (NMR)

form

solid

color

white to off-white

mp

245 °C (lit.)

solubility

acetonitrile: soluble

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

wet ice

storage temp.

−20°C

SMILES string

[I-].CC[N+](C)(CC)CCC1=C(C)c2ccc(OC)cc2OC1=O

InChI

1S/C18H26NO3.HI/c1-6-19(4,7-2)11-10-16-13(3)15-9-8-14(21-5)12-17(15)22-18(16)20;/h8-9,12H,6-7,10-11H2,1-5H3;1H/q+1;/p-1

InChI key

OAQIUABCTPFMOC-UHFFFAOYSA-M

Application

AMMC is demethylated by Human CYP2D6 to the fluorescent product 3-[2-(N,N-diethyl-N-methylammonium)ethyl]-hydroxy-4-methylcoumarin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

A1352-VAR:
A1352-25MG:
A1352-BULK:
A1352-5MG:4548173996158
A1352-5MG-PW:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service