Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

143R-1

Sigma-Aldrich

CD43 (SP55) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP55, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (143R-14)
vial of 0.5 mL concentrate (143R-15)
bottle of 1.0 mL predilute (143R-17)
vial of 1.0 mL concentrate (143R-16)
bottle of 7.0 mL predilute (143R-18)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... SPN(6693)

General description

CD43, also called leukosialin, is expressed on the surface of all leukocytes except for some resting B-cells. It appears to function as an anti-adhesion molecule, mediating repulsion amongst leukocytes. CD43 expression in lymphomas is highly correlated with CD5; thus most T-cell malignancies and a group of small lymphocyte B-cell malignancies [CLL/ SLL, mantle cell lymphoma (MCL), prolymphocytic leukemia] are often positive, while follicular lymphoma (FL) is usually negative. Anti-CD43 is less useful as a lineage marker, but is a sensitive indicator of aberrant B-cell populations. From 70 to 90% of T-cell lymphomas are labeled with this antibody. Plasmacytoma/myeloma and mast cell disease are also labeled with this antibody. No reactivity has been observed with reactive B-cells.

Quality


IVD

IVD

IVD

RUO

Linkage

CD43 Positive Control Slides, Product No. 143S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

143R-15:
143R-18:
143R-17:
143R-14:
143R-16:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnostic Immunohistochemistry: Theranostic and Genomic Applications (2006)
Manual of Diagnostic Antibodies for Immunohistology (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service