Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

62531

Sigma-Aldrich

Lithium hydroxide monohydrate

puriss. p.a., ≥99.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOH · H2O
CAS Number:
Molecular Weight:
41.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99.0% (T)
form:
powder or crystals

grade

puriss. p.a.

Assay

≥99.0% (T)

form

powder or crystals

pH

12 (0.4 g/L)

anion traces

chloride (Cl-): ≤50 mg/kg
lithium carbonate (Li2CO3): ≤1%
sulfate (SO42-): ≤100 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤20 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Li+].O.[OH-]

InChI

1S/Li.2H2O/h;2*1H2/q+1;;/p-1

InChI key

GLXDVVHUTZTUQK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hydroxide monohydrate is a hydrate of alkali metal hydroxide. Its IR spectral analysis has been reported. A refinement of its crystal structure has been described. Its crystals belong to the monoclinic crystal system having space group C2/m. It can be prepared by reacting Li metal with H2O, followed by the evaporation of the aqueous solution. It can be also be obtained from the basic aqueous solution of LiOH, via crystallization.

Application

Lithium hydroxide monohydrate (LiOH.H2O) may be employed as a catalyst for the melt transesterification of diphenyl carbonate and bisphenol A. Kinetics of this transesterification reaction have been investigated.

Other Notes

Reagent for the decomposition of oxides and silicates

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

62531-1KG-F:4548173940038
62531-VAR-F:
62531-250G-F:4548173940045
62531-VAR-F-D:
62531-BULK-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Hydrogen bonding. VI. Structural and infrared spectral analysis of lithium hydroxide monohydrate and cesium and rubidium hydroxide hydrates.
Gennick I and Harmon KM.
Inorganic Chemistry, 14(9), 2214-2219 (1975)
Infrared active vibrational modes of lithium hydroxide monohydrate.
Hase Y.
Inorganic and Nuclear Chemistry Letters, 16(3), 159-163 (1980)
Kinetics of melt transesterification of diphenyl carbonate and bisphenol A to polycarbonate with lithium hydroxide monohydrate catalyst.
Kim Y, et al.
Industrial & Engineering Chemistry Research, 31(9), 2118-2127 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service