Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

292591

Sigma-Aldrich

Activated charcoal

greener alternative

4-14 mesh, granular, Norit® PK 3-5

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52

vapor pressure

<0.1 mmHg ( 20 °C)

form

granular

autoignition temp.

842 °F

manufacturer/tradename

Norit® PK 3-5

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

particle size

4-14 mesh

mp

3550 °C (lit.)

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated Carbon (4-14 mesh, granular) can be used as a substrate or as an adsorbent to study the adsorption kinetics and characteristic of ethylenediaminetetraacetic acid (EDTA) and cadmium chelates.

Legal Information

Norit® is a registered trademark of Norit N.V.
Norit is a registered trademark of Norit N.V.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

292591-VAR:
292591-1KG:
292591-250G:
292591-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of the surface oxidation of activated carbon, the solution pH and the temperature on adsorption of ibuprofen
Guedidi H, et al.
Carbon, 54, 432-443 (2013)
Kishore Baathulaa et al.
Nature protocols, 6(12), 1990-1997 (2011-12-03)
Environment-sensitive fluorophores have different quantum yields in different solvents. 6-Dimethylaminonaphthalimide (6-DMN), for example, has a low quantum yield in aqueous solutions, but is highly fluorescent in nonpolar solvents or when bound to hydrophobic sites in proteins or membranes. 6-DMN has
Diazepam stability in wastewater and removal by advanced membranes technology, activated carbon and micelle-clay complex
Bufo SA, et al.
Desalination and Water Treatment (2015)
Activated carbon for gas separation and storage
Sircar S, et al.
Carbon, 34(1), 1-12 (1996)
About reactions occurring during chemical activation with hydroxides.
Lillo-Rodenas MA, et al.
Carbon, 42(7), 1371-1375 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service