D9809
Diatrizoic acid dihydrate
meets USP testing specifications
Synonym(s):
3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid dihydrate, Amidotrizoic acid dihydrate
About This Item
Recommended Products
Agency
USP/NF
meets USP testing specifications
Quality Level
Assay
98.0-102.0% anhydrous basis
form
solid
application(s)
hematology
histology
pharmaceutical (small molecule)
storage temp.
room temp
SMILES string
CC(=O)Nc1c(I)c(NC(C)=O)c(I)c(C(O)=O)c1I
InChI
1S/C11H9I3N2O4/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18/h1-2H3,(H,15,17)(H,16,18)(H,19,20)
InChI key
YVPYQUNUQOZFHG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
D9809-BULK:
D9809-VAR:
D9809-100G:
D9809-10G:4548173996394
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service