Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

73146

Supelco

1,3-Dipalmitoyl-2-oleoylglycerol

analytical standard

Synonym(s):

1,3-Dihexadecanoyl-2-(cis-9-octadecenoyl)glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H100O6
CAS Number:
Molecular Weight:
833.36
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (TLC)

shelf life

limited shelf life, expiry date on the label

composition

oleic acid, 33-36%
palmitic acid, 64-67%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

ester

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-32-35-38-41-44-47-53(56)59-50(48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2)49-58-52(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3/h25-26,50H,4-24,27-49H2,1-3H3/b26-25-

InChI key

FDCOHGHEADZEGF-QPLCGJKRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

73146-BULK:
73146-VAR:
73146-10MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service