Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

61828

Supelco

10-Undecen-1-ol

analytical standard

Synonym(s):

11-Hydroxy-1-undecene, NSC 66157, Undecylenyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)9OH
CAS Number:
Molecular Weight:
170.29
Beilstein:
1753225
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

refractive index

n20/D 1.448-1.453
n20/D 1.45 (lit.)

bp

132-133 °C/15 mmHg (lit.)
245-248 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCCCCC=C

InChI

1S/C11H22O/c1-2-3-4-5-6-7-8-9-10-11-12/h2,12H,1,3-11H2

InChI key

GIEMHYCMBGELGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

61828-1ML:
61828-BULK:
61828-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service