Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

42327

Supelco

Ethyl pyruvate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOOC2H5
CAS Number:
Molecular Weight:
116.12
Beilstein:
1071466
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.404 (lit.)

bp

144 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)C(C)=O

InChI

1S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3

InChI key

XXRCUYVCPSWGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl pyruvate is a volatile aliphatic ester of the pyruvate metabolite. It is present as a food additive/odor-active component in various food and beverage products. It is also used as an inhibitory odorant to repel the insects from human proximity.

Application

Ethyl pyruvate may be used as an analytical standard for the determination of the analyte in wine samples, and distilled spirits by gas chromatography (GC) based techniques.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

42327-1ML:
42327-BULK:
42327-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of evaporated ethyl pyruvate on reducing salmonella enteritidis in raw chicken meat
Cetin B, et al.
Brazilian Journal of Poultry Science, 21(2), 158-164 (2019)
Aromatic evolution of wine packed in virgin and recycled PET bottles
Dombre C, et al.
Food Chemistry, 176(2), 376-387 (2015)
Raspberry (Rubus idaeus L.) wine: Yeast selection, sensory evaluation and instrumental analysis of volatile and other compounds
Duarte WF, et al.
Food Research International, 43(9), 2303-2314 (2010)
Characterization of tequila flavor by instrumental and sensory analysis.
Benn SM & Peppard TL.
Journal of Agricultural and Food Chemistry, 44(2), 557-566 (1996)
Mitchell P Fink
Current opinion in anaesthesiology, 21(2), 160-167 (2008-04-30)
Ethyl pyruvate is a simple derivative of the endogenous metabolite, pyruvic acid. Ethyl pyruvate was shown to ameliorate some of the structural and functional changes associated with mesenteric ischemia and reperfusion in rats. Treatment with ethyl pyruvate has been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service