Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

37883

Supelco

1,4-Dimethylnaphthalene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH3)2
CAS Number:
Molecular Weight:
156.22
Beilstein:
2039842
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.613 (lit.)

bp

262-264 °C/751 mmHg (lit.)

mp

−18 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1ccc(C)c2ccccc12

InChI

1S/C12H12/c1-9-7-8-10(2)12-6-4-3-5-11(9)12/h3-8H,1-2H3

InChI key

APQSQLNWAIULLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

37883-BULK:
37883-100MG:
37883-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damir Posavec et al.
Organic & biomolecular chemistry, 10(35), 7062-7069 (2012-08-01)
Decomposition of endoperoxide containing molecules is an attractive approach for the delayed release of singlet oxygen under mild reaction conditions. Here we describe a new method for the adaptation of the corresponding decay times by controlling the supramolecular functional structure
Michael A Campbell et al.
Functional & integrative genomics, 12(3), 533-541 (2011-11-25)
The suppression of sprout growth is critical for the long-term storage of potato tubers. 1,4-Dimethylenapthlene (DMN) is a new class of sprout control agent but the metabolic mode of action for this compound has yet to be elucidated. Changes in
D S Malament et al.
Carcinogenesis, 2(8), 723-729 (1981-01-01)
Phenanthrene (Phe) and to a lesser degree 1,4-dimethylnaphthalene (DMeN) were each found to retard the development of epidermoid carcinomas in hamster buccal pouch induced by the thrice weekly application of a 0.5 per cent solution of 7,12-dimethylbenz[a]anthracene (DMBA) in heavy
A Kilanowicz et al.
International journal of occupational medicine and environmental health, 13(4), 325-334 (2001-03-30)
The distribution, excretion and metabolism of 1,4-dimethylnaphthalene following i.p. administration of a single dose of 28 mg/kg to rats, was investigated using radiotracer [3H] and gas chromatography-mass spectrometry technique (GC-MS). After 72 h, about 97% of the given dose was
P Di Mascio et al.
Biological chemistry Hoppe-Seyler, 376(5), 297-301 (1995-05-01)
Carotenoids comprise one of the most widespread classes of pigments found in nature. Polyene pigments from the dinoflagellate Gonyaulax polyedra were extracted every hour over a 24 hour period and the levels of beta-carotene during the day-phase were found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service