Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

16382

Supelco

Casticin

analytical standard

Synonym(s):

3′,5-Dihydroxy-3,4′,6,7-tetramethoxyflavone, 3,4′,6,7-Tetra-O-methylquercetagetin, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxy-4H-1-benzopyran-4-one, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxychromen-4-one, Quercetagetin 3,4′,6,7-tetramethyl ether, Vitexicarpin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O8
CAS Number:
Molecular Weight:
374.34
Beilstein:
1300392
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)C2=C(OC)C(=O)c3c(O)c(OC)c(OC)cc3O2

InChI

1S/C19H18O8/c1-23-11-6-5-9(7-10(11)20)17-19(26-4)16(22)14-12(27-17)8-13(24-2)18(25-3)15(14)21/h5-8,20-21H,1-4H3

InChI key

PJQLSMYMOKWUJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Casticin is a flavonoid, widely used as an anti-inflammatory agent. It has anti-tumor properties and is considered to be a potent immunomodulatory and cytotoxic compound.

Application

It was may be used as reference standard to compare retention time and ion peaks in plant extracts during GC/MS analysis to quantify Casticin and artemetin.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: achillea

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

16382-VAR:
16382-10MG:
16382-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanxiang Zeng et al.
Acta biochimica et biophysica Sinica, 44(5), 442-449 (2012-03-20)
Casticin, a polymethoxyflavone from Fructus viticis used as an anti-inflammatory agent in Chinese traditional medicine, has been reported to have anti-cancer activity. The purpose of this study was to examine the apoptotic activity of casticin on human cervical cancer cells
Duck-jae Koh et al.
Journal of ethnopharmacology, 136(3), 399-405 (2011-01-22)
The fruits of Vitex rotundifolia L. have long been used for the treatment of inflammation of the respiratory tract in East Asia. To determine if casticin, one of the constituents of Vitex rotundifolia L., has anti-allergic and anti-inflammatory effects in
Yun Ling et al.
Biomedical chromatography : BMC, 26(12), 1502-1508 (2012-03-13)
Casticin (3',5-dihydroxy-3, 4',6,7-tetramethoxyflavone) has been revealed to possess various kinds of pharmacological activities, including immunomodulatory, anti-hyperprolactinemia, anti-tumor and neuroprotetective activities. In order to gain an understanding of the biotransformation of casticin in vivo, a systematic method based on liquid chromatography-electrospray
Xiao Han et al.
Journal of chromatography. A, 1151(1-2), 180-182 (2007-03-22)
Following an initial clean-up step on silica, high-speed counter-current chromatography (HSCCC) was used to purify a flavone, casticin (5,3'-dihydroxy-3,6,7, 4'-tetramethoxyflavone), from an extract of the dried leaves of Artemisia annua L. The two-phase solvent system used was composed of n-hexane-ethyl
Qi YE et al.
Acta pharmacologica Sinica, 31(12), 1564-1568 (2010-11-03)
to investigate the anti-hyperprolactinemia activity of casticin, a flavonoid isolated from Vitex rotundifolia, and elucidate its molecular mechanism. hyperprolactinemia (MIHP) was induced by administration of metoclopramide dihydrochloride (50 mg/kg, tid, ip, for 10 d) in SD rats and the primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service