Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

14515

Supelco

Vanillic acid 4-β-D-glucoside

analytical standard

Synonym(s):

4-Hydroxy-3-methoxybenzoic acid 4-β-D-glucoside, Vanillic acid 4-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O9
CAS Number:
Molecular Weight:
330.29
Beilstein:
5452207
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water

application(s)

food and beverages

format

neat

SMILES string

COc1cc(ccc1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C(O)=O

InChI

1S/C14H18O9/c1-21-8-4-6(13(19)20)2-3-7(8)22-14-12(18)11(17)10(16)9(5-15)23-14/h2-4,9-12,14-18H,5H2,1H3,(H,19,20)/t9-,10-,11+,12-,14-/m1/s1

InChI key

JYFOSWJYZIVJPO-YGEZULPYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

14515-VAR:
14515-BULK:
14515-5MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service