Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-5710

Sigma-Aldrich

Sulfamic acid

SAJ first grade, ≥99.0%

Synonym(s):

Amidosulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2SO3H
CAS Number:
Molecular Weight:
97.09
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

215-225 °C (dec.) (lit.)

density

2.151 g/cm3 at 25 °C

SMILES string

NS(O)(=O)=O

InChI

1S/H3NO3S/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

IIACRCGMVDHOTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

28-5710-5-500G-J:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Yves Winum et al.
Bioorganic & medicinal chemistry, 21(6), 1419-1426 (2012-12-04)
Targeting tumour associated carbonic anhydrase (CA, EC 4.2.1.1) isoforms IX and XII is now considered as a pertinent approach for the development of new cancer therapeutics against hypoxic tumours. In the last period, with the help of X-ray crystallography, much
Patricia Chávez et al.
The Journal of organic chemistry, 77(4), 1922-1930 (2012-01-31)
An intramolecular diamination of acrylates is reported using sulfamates as nitrogen sources. This reaction proceeds under palladium(II) catalysis with copper bromide as oxidant and gives rise to anti-configured 2,3-diamino carboxylates as bicyclic sulfamate derivatives. An aminobrominated intermediate within the diamination
Na Zhang et al.
The Journal of organic chemistry, 77(14), 5956-5964 (2012-06-21)
The efficiency of arylboron-based nucleophiles, boronic acid, potassium trifluoroborate, neopentylglycolboronate, and pinacol boronate in nickel-catalyzed Suzuki-Miyaura cross-coupling reactions with the two C-O electrophiles, mesylates, and sulfamates was compared. Arylboronic acid is the most reactive and most atom-economic of the four
Amit Saha et al.
Chemical communications (Cambridge, England), 48(71), 8889-8891 (2012-07-21)
Unnatural α-amino acids containing dithiocarbamate side chains were synthesized by a one-pot reaction of in situ generated dithiocarbamate anions with sulfamidates. A wide range of these anions participated in the highly regio- and stereo-selective ring opening of sulfamidates to produce
Derek J Langeslay et al.
Analytical chemistry, 83(20), 8006-8010 (2011-09-15)
Sulfamate (NHSO(3)(-)) groups are critically important structural elements of the glycosaminoglycans heparin and heparan sulfate (HS). Experimental conditions are presented for detection of the sulfamate (1)H NMR resonances in aqueous solution. NMR spectra reported for N-sulfoglucosamine (GlcNS) and the synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service