Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-4510

Sigma-Aldrich

Sodium thiosulfate pentahydrate

JIS special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
grade:
JIS special grade
form:
crystalline

grade

JIS special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

28-4510-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael C Reade et al.
Emergency medicine Australasia : EMA, 24(3), 225-238 (2012-06-08)
Cyanide poisoning is uncommon, but generates interest because of the presumed utility of an antidote immediately available in those areas with a high risk of cyanide exposure. As part of its regular review of guidelines, the Australian Resuscitation Council conducted
Steven B Dethloff
Nephrology nursing journal : journal of the American Nephrology Nurses' Association, 39(4), 323-325 (2012-10-16)
Though CUA therapy with IP sodium thiosulfate represents an off-label usage of the drug, reports of its successful use are on the rise. It continues to be successfully administered both IP and IV, and in adult and pediatric populations. The
Vikhyat S Bebarta et al.
Annals of emergency medicine, 59(6), 532-539 (2012-03-06)
We compare the efficacy of hydroxocobalamin to sodium thiosulfate to reverse the depressive effects on mean arterial pressure in a swine model of acute cyanide toxicity and gain a better understanding of the mechanism of action of the hydroxocobalamin in
Fengbo Xie et al.
The Analyst, 137(6), 1343-1350 (2012-01-05)
Nephrotoxicity is one of the major concerns for anticancer drug safety because most drugs are metabolized and excreted by the kidneys. Convenient tools able to perform rapid in vitro cytotoxicity analysis and identify drug side effects in kidney cells during
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 4(9), 960-967 (2012-07-31)
The anticancer drug cis-platin (CP) is widely used to treat patients, but it is also associated with significant side effects, including nephrotoxicity. Given that this metallodrug is intravenously (iv) administered, its biotransformations in the bloodstream are likely to be involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service