Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-4300

Sigma-Aldrich

Sodium sulfide nonahydrate

SAJ first grade, 90.0-102.0%, storage temp.:2-8°C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S·9H2O
CAS Number:
Molecular Weight:
240.18
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
grade:
SAJ first grade
form:
crystalline

grade

SAJ first grade

Assay

90.0-102.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: sodium

availability

available only in Japan

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.O.O.[Na]S[Na]

InChI

1S/2Na.9H2O.S/h;;9*1H2;

InChI key

UIINQVYGFKNSPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

28-4300-5-500G-J:
28-4300-8-18KG-J:
28-4300-8-20KG-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renfa Liu et al.
Theranostics, 5(10), 1144-1153 (2015-08-19)
The integration of diagnostic and therapeutic functionalities into one nanoplatform shows great promise in cancer therapy. In this research, manganese (II) chelate functionalized copper sulfide nanoparticles were successfully prepared using a facile hydrothermal method. The obtained ultrasmall nanoparticles exhibit excellent
Danielle E Gorka et al.
Environmental science & technology, 49(16), 10093-10098 (2015-07-07)
The use of antibacterial silver nanomaterials in consumer products ranging from textiles to toys has given rise to concerns over their environmental toxicity. These materials, primarily nanoparticles, have been shown to be toxic to a wide range of organisms; thus
Shin Koike et al.
Biochemical and biophysical research communications, 459(3), 488-492 (2015-03-10)
Polysulfides are a typical type of bound sulfur, which is physiologically stable form of sulfur species, derived from the hydrogen sulfide (H2S) that is generated endogenously in cells. We previously reported that bound sulfur protects neuronal cells from oxidative injury.
Josephine Selvaraj et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(10), 2010-2018 (2014-07-01)
Nitrogen (N)-doped reduced graphene oxide (nRGO) is systematically incorporated into a TiO(2) -CdS photoelectrochemical (PEC) cell and its role is examined in the three main components of the cell: 1) the CdS-sensitized TiO(2) photoanode, 2) the cathode, and 3) the
Hongxia Li et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(10), 1011-1017 (2015-10-16)
This paper describes a convenient and rapid fluorescence sensor for determination of paraquat (PA) based on glutathione-capped CdS quantum dots (QDs). The methodology enabled the use of a simple synthesis procedure for water solubilization of CdS QDs via a fast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service