Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

15-0590

Sigma-Aldrich

Iodine

JIS special grade, ≥99.8%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
EC Number:
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:

grade

JIS special grade

vapor density

9 (vs air)

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

Assay

≥99.8%

availability

available only in Japan

resistivity

1.3E15 μΩ-cm

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

15-0590-2-25G-J:
15-0590-5-500G-J:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eike I Piechowiak et al.
Radiology, 275(3), 692-697 (2015-02-06)
To determine the effect of the use of iodinated contrast agents on the formation of DNA double-strand breaks during chest computed tomography (CT). This study was approved by the institutional review board, and written informed consent was obtained from all
Michael B Zimmermann
Thyroid : official journal of the American Thyroid Association, 17(9), 829-835 (2007-10-25)
Iodine is required for the production of thyroid hormones, which are essential for normal brain development, and the fetus, newborn, and young child are particularly vulnerable to iodine deficiency. The iodine requirement increases during pregnancy and recommended intakes are in
Zulfiqar A Bhutta et al.
Lancet (London, England), 382(9890), 452-477 (2013-06-12)
Maternal undernutrition contributes to 800,000 neonatal deaths annually through small for gestational age births; stunting, wasting, and micronutrient deficiencies are estimated to underlie nearly 3·1 million child deaths annually. Progress has been made with many interventions implemented at scale and
Michael B Zimmermann et al.
Nutrition reviews, 70(10), 553-570 (2012-10-06)
Iodine status has been historically assessed by palpation of the thyroid and reported as goiter rates. Goiter is a functional biomarker that can be applied to both individuals and populations, but it is subjective. Iodine status is now assessed using
Peter N Taylor et al.
The Journal of clinical endocrinology and metabolism, 99(11), 4291-4298 (2014-07-25)
Thyroid dysfunction is associated with impaired cognitive development. Perchlorate decreases thyroidal iodine uptake, potentially reducing thyroid hormone production. It is unclear whether perchlorate exposure in early life affects neurodevelopment. Historical cohort analysis. From 2002 to 2006, 21,846 women at gestational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service