03-1565
Benzoyl peroxide
SAJ first grade, ≥98.0% dry basis
Synonym(s):
Dibenzoyl peroxide
About This Item
Recommended Products
grade
SAJ first grade
Assay
≥98.0% dry basis
form
solid
autoignition temp.
176 °F
contains
~25% water as stabilizer
reaction suitability
reagent type: oxidant
availability
available only in Japan
mp
105 °C (lit.)
SMILES string
O=C(OOC(=O)c1ccccc1)c2ccccc2
InChI
1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H
InChI key
OMPJBNCRMGITSC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Org. Perox. B - Skin Sens. 1A
Storage Class Code
4.1A - Other explosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
FSL
Group 5: Self-reactive substances
Organic peroxides
Hazardous rank I
1st self-reactive materials
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
03-1565-6-100G-J:
03-1565-5-500G-J:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service