01-4268
Ammonia Nitrogen Standard Solution
1000 ppm
Synonym(s):
Ammonium chloride solution
About This Item
Recommended Products
form
liquid
availability
available only in Japan
concentration
1000 ppm
pH
5.7 (23.2 °C)
density
0.995 g/mL at 25 °C (lit.)
SMILES string
[Cl-].[H][N+]([H])([H])[H]
InChI
1S/ClH.H3N/h1H;1H3
InChI key
NLXLAEXVIDQMFP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
01-4268-6-100ML-J:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service