Skip to Content
Merck
All Photos(1)

Key Documents

HG000C96

Millipore

Cetrimide Agar Base

pH 7.2±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111

composition

Agar, 13.6 g/L
Cetrimide, 0.5 g/L
Magnesium Chloride, 1.4 g/L
Peptone, 20.5 g/L
Potassium Sulphate, 10.0 g/L

packaging

poly bottle of 12 × 500 g (HG000C96.500)

pH

7.2±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

A selective agar for the isolation and identification of Pseudomonas aeruginosa from various materials.

Preparation Note

Suspend 46g in 1 litre demineralised water. Add 10ml glycerol. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes. Cool to 45 - 50°C, mix well and pour plates.
Incubate at 37°C for 48 hours

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service