EZHAWG47BW
EZ-Pak® Membrane Filter
EZ-Pak®, filter 0.45 μm, filter diam. 47 mm, circles white mixed cellulose esters (MCE) filter (gridded)
Synonym(s):
MCE Membrane, Membrane Filter, Mixed Cellulose Filter
About This Item
Recommended Products
material
circles white mixed cellulose esters (MCE) filter (gridded)
Quality Level
Agency
ISO 7704
sterility
sterile; ethylene oxide treated
manufacturer/tradename
EZ-Pak®
filter diam.
47 mm
pore size
0.45 μm filter
application(s)
food and beverages
water monitoring
shipped in
ambient
General description
After the Sample filtration the EZ-Pak® Membrane is incubated together with a selected culture media which will lead to the recovery of the targeted microorganism.
Application
Features and Benefits
- Ready to use.
- Sterile.
- Continuous Base and Individually Packed.
- Higher Recoveries.
- Convenient Handling.
- Minimal Cross Contamination.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PRTR
Class I Designated Chemical Substances
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
EZHAWG47BW:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service