Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

AB2295

Sigma-Aldrich

Anti-phospho GluR4 (Ser842) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

glutamate receptor, AMPA 4, Glutamate receptor ionotropic, AMPA 4, AMPA-selective glutamate receptor 4, glutamate receptor 4, GluR-D, GluR-4, Glutamate receptor 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
rat
technique(s):
western blot: suitable
citations:

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer842)

Gene Information

human ... GRIA4(2893)

General description

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. GluR4 is an ionotropic glutamate receptor encoded by the GRIA4 gene. Alternative splicings of this gene result in transcript variants encoding different isoforms, which are believed to vary in their signal transduction properties. Some haplotypes of this gene show a positive association with schizophrenia. Direct phosphorylation of GluR4-containing receptors may regulate rapid excitatory synaptic transmission. In particular, phosphorylation of GluR4 at Serine 842 is both necessary and sufficient to deliver the receptor to the synapse and induce plasticity in early postnatal development.

Specificity

This antibody recognizes GluR4 when phosphorylated at Ser842.

Immunogen

Epitope: Phosphorylated Ser842
Linear peptide corresponding to rat GluR4 phosphorylated at Ser842.

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-phospho GluR4 (Ser842) Antibody is validated for use in WB for the detection of phospho GluR4 (Ser842).

Quality

Evaluated by Western Blot in phosphatase treated and untreated rat cerebellum tissue lysates.

Western Blot Analysis: 0.5 µg/mL of this antibody detected GluR4 on 10 µg of phosphatase treated and untreated rat cerebellum tissue lysates.

Target description

~101 kDa observed. An uncharacterized band may be visible at ~39 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Phosphatase treated and untreated rat cerebellum tissue lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AB2295:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service