Skip to Content
Merck
All Photos(1)

Key Documents

9.43442

Supelco

Combi vessel, 10 l

metal, PE liner, blue, lacquered, 2 in. (coarse threads)

Synonym(s):

10 l metal drum steel/PE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24112108

material

PE liner
metal

color

blue, lacquered

fitting

2 in. (coarse threads)

General description

We offer a big variety of packaging materials like empty bottles and containers for your own preparations and mixtures of our reagents.
metal drum with PE liner, lacquered blue, with 2" coarse thread

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service