8.16104
(R,S)-(+,-)-4-Methyloctanoic acid
for synthesis
Synonym(s):
(R,S)-(+,-)-4-Methyloctanoic acid
About This Item
Recommended Products
Quality Level
Assay
≥98.0% (acidimetric)
form
liquid
bp
292-293 °C
transition temp
flash point >99 °C
density
0.904-0.914 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/C9H18O2/c1-3-4-5-8(2)6-7-9(10)11/h8H,3-7H2,1-2H3,(H,10,11)/p-1/t8-/m1/s1
InChI key
LEGGANXCVQPIAI-MRVPVSSYSA-M
Analysis Note
Density (d 20 °C/ 4 °C): 0.904 - 0.914
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
FSL
Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service