Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

528884

Sigma-Aldrich

Polo-like Kinase Inhibitor V, Poloxin

The Polo-like Kinase Inhibitor V, Poloxin, also referenced under CAS 321688-88-4, controls the biological activity of Polo-like Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Polo-like Kinase Inhibitor V, Poloxin, Polo-like Kinase Inhibitor V, 2-isopropyl-5-methyl-4-(2-methylbenzoyloxyimino)cyclohexa-2,5-dienone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO3
CAS Number:
Molecular Weight:
297.35
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL, clear, yellow

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H19NO3/c1-11(2)15-10-16(13(4)9-17(15)20)19-22-18(21)14-8-6-5-7-12(14)3/h5-11H,1-4H3/b19-16-

InChI key

CMOJHDQJJPIVEC-MNDPQUGUSA-N

General description

A thymoquinone derivative, non-ATP competitive Polo-like Kinase 1 (Plk1) inhibitor specifically targeting the polo-box domain (PBD) of Plk1 (IC50) = 4.8 µM. Reduces both chromosome and kinetochoral localization of Plk1, induces chromosome congression defects , mitotic arrest in prometaphase, and apoptosis in HeLa cells at 25 µM. Exhibits anti-proliferative effects in a range of cancer cell lines, and in a mouse Xenograft model involving HeLa and MDA-MB-231 cells at 40 mg/kg.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Reindl, W., et al. 2008. Chem Biol.15, 459.
Yuan, J., et al. 2011, Am J Pathol179, 2091.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

528884-MG:
528884-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service