Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

500587

Sigma-Aldrich

γ-Secretase Inhibitor XXV, FT-9

Synonym(s):

γ-Secretase Inhibitor XXV, FT-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12Cl2O3
Molecular Weight:
311.16
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

FT-9, a cell-permeable NSAID-like compound, is a partially selective γ-Secretase Inhibitor in both microsomes and whole cells. Displays dose-dependent inhibition of the γ-secretase cleavage of β-Amyloid Precursor Protein (APP) and APLP2 with IC50 values against APP ICD (intracellular domain) and APLP2 ICD of ˜260 µM, but there is a less potent effect on γ-secretase processing of APLP1. The concentration of FT-9 required to cause a 2-fold increase in APP CTFs, 3.4 +/- 0.4 µM, is significantly lower than that for APLP1 (25.7 +/- 8.9 µM) (P = 0.001). For comparison, the concentration of another γ-Secretase inhibitor compound E (EMD Chemicals, Inc. Cat. No. 565790) needed for a 2-fold increase in the magnitude of the CTF band signal is not significantly different, being 0.27 nM for APP versus 0.24 nM for APLP1. This is the first example that a GSI can preferentially target APP processing while sparing other γ-substrates.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sala Frigerio, C., et al. 2009. Biochemistry48, 10894.
Kukar, TL., et al. 2008. Nature453, 925.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

500587-10MG:
500587-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service