Skip to Content
Merck
All Photos(1)

Key Documents

444145

Sigma-Aldrich

MDM2 Inhibitor

The MDM2 Inhibitor, also referenced under CAS 562823-84-1, controls the biological activity of MDM2.

Synonym(s):

MDM2 Inhibitor, trans-4-Iodo, 4ʹ-boranyl-chalcone, MDM2 Antagonist IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12BIO3
CAS Number:
Molecular Weight:
377.97
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H12BIO3/c17-14-8-1-11(2-9-14)3-10-15(18)12-4-6-13(7-5-12)16(19)20/h1-10,19-20H/b10-3+

InChI key

BYMGWCQXSPGCMW-XCVCLJGOSA-N

General description

A cell-permeable boranyl-chalcone compound that binds strongly to MDM2 and irreversibly disrupts MDM2/p53 protein complex formation. Exhibits selective toxicity to MDM2-overexpressing human breast cancer cell lines (IC50 = 10 µM for MDA-MB-435, 8.8 µM for MDA-MB-231, and 7 µM for Wt-MCF7) compared to normal breast cell lines (IC50 = 75 µM for MCF-10A and 63 µM for MCF-12A).
A cell-permeable boranyl-chalcone that binds strongly to MDM2 and irreversibly disrupts MDM2/p53 protein complex. Exhibits selective toxicity towards MDM2 overexpressing human breast cancer cell lines (IC50 = 10, 8.8, and 7 µM for MDA-MB-435, MDA-MB-231, and Wt-MCF7, respectively) compared to normal breast cell lines (IC50 = 75 and 63 µM for MCF-10A and MCF-12A, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MDM2
Product does not compete with ATP.
Reversible: no
Target IC50: 10, 8.8, and 7 µM in selective toxicity for MDA-MB-435, MDA-MB-231, and Wt-MCF7, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kumar, S.K., et al. 2003. J. Med. Chem.46, 2813.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srinivas K Kumar et al.
Journal of medicinal chemistry, 46(14), 2813-2815 (2003-06-27)
A series of boronic-chalcone derivatives were synthesized and tested for antitumor activity against human breast cancer cell lines. The results show the boronic-chalcones are more toxic to breast cancer cells compared to normal breast cells than other known chalcones.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service