Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

361568

Sigma-Aldrich

GSK-3β Inhibitor XXV

The GSK-3β Inhibitor XXV controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor XXV, 5-(4-Nitrophenyl)-N-(3-chloro-4-methyl)phenyl-1,3,4-oxadiazol-2-amine, N-(3-Chloro-4-methylphenyl)-5-(4-nitrophenyl)-1,3,4-oxadiazol-2-amine, M SEW00923SC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClN4O3
CAS Number:
Molecular Weight:
330.73
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C=C1)=C(Cl)C=C1NC2=NN=C(C3=CC=C(C=C3)[N+]([O-])=O)O2

General description

A blood brain barrier permeable oxadiazole compound that acts as a potent, reversible and ATP-competitive inhibitor of GSK-3β activity (IC50 = 17.1 nM) with excellent selectivity over Cdk2 (22% inhibition at 10 µM). Shown to elevate liver glycogen levels by ~3.5- and ~4.3-fold in C57BL/6N mice when dosed at 5 and 15 mg/kg (i.p.) amounts.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Khanfar, M.A., et al. 2010. J. Med. Chem.53, 8534.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

361568-MG:
361568-10MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service