Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

356780

Millipore

Glycyrrhizin, Ammonium Salt

Inhibits replication, adsorption, and penetration of severe acute respiratory syndrome-associated coronavirus (SARS-CV). It is allso a known 11β-hydroxysteroid dehydrogenase type 2 (11-HSD2) inhibitor.

Synonym(s):

Glycyrrhizin, Ammonium Salt, Glycyrrhizic Acid, NH₄ +

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H61O16 · NH4
CAS Number:
Molecular Weight:
839.96
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

aqueous NH4OH: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C42H62O16.H3N/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50;/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54);1H3/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+;/m0./s1

InChI key

ILRKKHJEINIICQ-OOFFSTKBSA-N

General description

A triterpenoid saponin isolated from liquorice roots that displays antiviral and anti-tumor activities. Inhibits replication, adsorption, and penetration of severe acute respiratory syndrome-associated coronavirus (SARS-CV), presumably via its ability to upregulate nitric oxide synthase activity. Reported to block the growth of other viruses, including herpes virus, and helps restore liver functions in patients with hepatitis C. It is allso a known 11β-hydroxysteroid dehydrogenase type 2 (11-HSD2) inhibitor.
A triterpenoid saponin, isolated from liquorice roots, that exhibits anti-viral and anti-tumor activities. Inhibits replication, adsorption, and penetration of severe acute respiratory syndrome-associated coronavirus (SARS-CV), presumably as a result of its ability to upregulate nitric oxide synthase activity. Also reported to hamper the growth of other viruses, including herpes virus. Shown to restore liver function in patients with hepatitis C virus (HCV). Also shown to inhibit 11β-hydroxysteroid dehydrogenase type 2 (11-HSD2).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cinatl, J., et al. 2003. Lancet361, 2045.
Tanahashi, T., et al. 2002. J. Steroid Biochem. Mol. Biol.80, 441.
Kondo, Y., and Takano, F. 1994. Biol. Pharm. Bull.17, 759.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

356780-5GM:
356780-GM:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service