Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

324745

Sigma-Aldrich

Elastase Inhibitor III

The Elastase Inhibitor III controls the biological activity of Elastase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Elastase Inhibitor III, HLE Inhibitor, MeOSuc-AAPV-CMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H35ClN4O7
Molecular Weight:
502.99
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 10 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

−20°C

General description

A potent inhibitor of human leukocyte elastase (HLE) (Ki = 10 µM).
A potent irreversible inhibitor of human leukocyte elastase (HLE)(Ki = 10 µM). Protects against lung injury induced by instillation of elastase.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Human leukocyte elastase (HLE)
Product does not compete with ATP.
Reversible: no
Target Ki: 10 µM against human leukocyte elastase (HLE)

Warning

Toxicity: Standard Handling (A)

Sequence

MeOSuc-Ala-Ala-Pro-Val-CMK

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fletcher, D.S., et al. 1990. Am. Rev. Respir. Dis.141, 672.
Stein, R.L., and Trainor, D.A. 1986. Biochemistry 25, 5414.
Powers, J.C., et al. 1977. Biochim. Biophys. Acta 485, 15.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

324745-5MG:
324745-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service