Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

252801

Sigma-Aldrich

DGAT-1 Inhibitor, A 922500

The DGAT-1 Inhibitor, A 922500, also referenced under CAS 959122-11-3, controls the biological activity of DGAT-1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

DGAT-1 Inhibitor, A 922500

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N2O4
CAS Number:
Molecular Weight:
428.48
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

light yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C26H24N2O4/c29-24(22-7-4-8-23(22)25(30)31)19-11-9-17(10-12-19)18-13-15-21(16-14-18)28-26(32)27-20-5-2-1-3-6-20/h1-3,5-6,9-16,22-23H,4,7-8H2,(H,30,31)(H2,27,28,32)/t22-,23-/m1/s1

InChI key

BOZRFEQDOFSZBV-DHIUTWEWSA-N

General description

An orally, bioavailable human and mouse diacylglycerol acyltransferase DGAT-1 inhibitor (IC50 = 9nM and 22 nM, respectively), which may target DGAT-2 (IC50 = 53 µM) and the phylogenetic family members acyl coenzyme A cholesterol acyltransferase-1 and -2 (IC50 = 296 µM) with equal or less potencies. In vivo activities indicate an effective reduction in serum triglycerides, FFA concentrations, and total cholesterol by 53%, 55% and 25%, respectively, compared to baseline, when administered orally at 3 mg/kg daily dosages in Dyslipidemic Hamsters. Similar reductions in triglyceride and FFA concentrations were shown in Zucker fatty rats.

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

King, A.J., et al. 2009. J. Pharmacol. Exp. Ther.330, 526.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

252801-MG:
252801-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service