Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

239823

Sigma-Aldrich

CXCR3 Agonist, PS372424

The CXCR3 Agonist, PS372424 controls the biological activity of CXCR3. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

CXCR3 Agonist, PS372424, (S)-N-((S)-1-((cyclohexylmethyl)amino)-5-guanidino-1-oxopentan-2-yl)-2-(4-oxo-4-phenylbutanoyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxamide, Chemokine (C-X-C motif) Receptor 3 Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H44N6O4 · 3H2O
CAS Number:
Molecular Weight:
642.79
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

C1CCC(CC1)CNC(=O)C(CCCN=C(N)N)NC(=O)C2CC3=CC=CC=C3CN2C(=O)CCC(=O)C4=CC=CC=C4

General description

A peptidomimetic derivative containing tetrahydroisoquinoline-arginine motif that aligns with a Pro-Arg motif of CXCL10. Acts as a highly specific agonist of human CXCR3 (EC50 = 1.1 mM, competitive CXCL10 binding IC50 = 42 nM). Does not affect murine CXCR3. Induces a long-lasting internalization of about 87% of cell-surface CXCR3 within 30 min. Effectively stimulates T-cell migration (~ 50 nM) and blocks the migration of activated T cells towards CXCL11, CXCL12, and CCL5. Also reduces the migration of activated T cells towards rheumatoid arthritis synovial fibroblasts (RASF). Shown to induce phosphorylation of ERK with similar potency and kinetics as CXCL11. Also shown to block CXCL11 induced migration of CD45+ cells to air pouches generated on humanized mice.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

O′Boyle, G., et al. 2012, PNAS.109, 4598.
Stroke, I. L., et al. 2006, Biochem Biophys Res Comm.349, 221.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

239823-MG:
239823-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service