Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

233630

Sigma-Aldrich

Cryptotanshinone

Cryptotanshinone is shown to inhibit the constitutive STAT3 Tyr⁷⁰⁵ phosphorylation in DU145 prostate cancer cells independent of the IL-6/JAK/STAT3 signaling pathway.

Synonym(s):

Cryptotanshinone, AMPK Signaling Activator VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20O3
CAS Number:
Molecular Weight:
296.36
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

ethanol: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C19H20O3/c1-10-9-22-18-12-6-7-13-11(5-4-8-19(13,2)3)15(12)17(21)16(20)14(10)18/h6-7,10H,4-5,8-9H2,1-3H3/t10-/m0/s1

InChI key

GVKKJJOMQCNPGB-JTQLQIEISA-N

General description

Cryptotanshinone, a cell-permeable diterpene quinone and naturally occurring herbal constituent of Salvia miltiorrhiza Bunge (Danshen), is shown to inhibit the constitutive STAT3 Tyr705 phosphorylation in DU145 prostate cancer cells (>90% inhibition in 1 h at 7 µM) independent of the IL-6/JAK/STAT3 signaling pathway, resulting in a blockage of STAT3 dimerization, nuclear translocation, and STAT3-dependent transcription activity (by ~80% in 24 h at 7 µM). Cryptotanshinone is also reported to deplete cellular ATP level (by 75% in 12 h at 20 µM in C2C12 myotubes), resulting in an indirect activation of cellular AMPK pathways and an enhancement of insulin-stimulated cellular glucose uptake in C2C12 myotubes and 3T3-L1 adipocytes (by ~1.3- and 1.9-fold, respectively). Cryptotanshinone is demonstrated to be an orally active hypoglycemic agent in ob/ob mice, db/db mice, and ZDF rats in vivo (600 mg/kg/day).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Shin, D.S., et al. 2009. Cancer Res.69, 193.
Kim, E.J., et al. 2007. Mol. Pharmacol.72, 62.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

233630-MG:
233630-1.1ML:
233630-10MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service