Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

200016

Sigma-Aldrich

Nα-Benzoyl-L-arginine Ethyl Ester, Hydrochloride

Substrate for bromelain, ficin, kallikrein, papain, subtilisin, thrombin, and trypsin.

Synonym(s):

Nα-Benzoyl-L-arginine Ethyl Ester, Hydrochloride, BAEE, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N4O3
CAS Number:
Molecular Weight:
306.36
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 25 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C15H22N4O3.ClH/c1-2-22-14(21)12(9-6-10-18-15(16)17)19-13(20)11-7-4-3-5-8-11;/h3-5,7-8,12H,2,6,9-10H2,1H3,(H,19,20)(H4,16,17,18);1H

InChI key

HIXDELXKSSLIKB-UHFFFAOYSA-N

General description

Substrate for bromelain, ficin, kallikrein, papain, subtilisin, thrombin, and trypsin.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for bromelain, ficin, kallikrein, papain, subtilisin, thrombin, and trypsin
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fadnavis, N.W., et al. 1998. Biochimie80, 1025.
Glazer, A.N. 1967. J. Biol. Chem. 242, 433.
Schwert, G., et al. 1955. Biochim. Biophys. Acta 16, 570.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

200016-GM:
200016-5GM:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service