Skip to Content
Merck
All Photos(1)

Key Documents

1.15072

Supelco

Phosphorus standard dissolved in oil

1 g/kg P Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

Quality Level

transition temp

flash point 63 °C

density

0.86 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Assay (P): 980 - 1020 mg/kg
standard deviation (s) (± mg/kg):
Determination method: HPLC vs. external Standard

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(C)

63 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service