Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

1.09736

Supelco

Methyl myristate

reference substance for gas chromatography

Synonym(s):

Methyl myristate, Myristic acid methyl ester, Methyl tetradecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOCH3
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-680-1

vapor pressure

0.065 Pa ( 25 °C)

Quality Level

form

liquid

bp

295 °C/1001 hPa

mp

18 °C

transition temp

flash point 112 °C

density

0.867 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C15H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h3-14H2,1-2H3

InChI key

ZAZKJZBWRNNLDS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112.0 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service