1.07565
Extran® AP 13
powder, alkaline with detergents
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
form
solid
pH
12.2 (20 °C, 3 g/L in H2O)
bulk density
850 kg/m3
storage temp.
no temp limit
Analysis Note
Identity: passes test
pH-value (1 %; water): ≥ 12.5
Foaming volume (5 %; water): ≤ 1 ml
Bulk density: about 85
pH-value (1 %; water): ≥ 12.5
Foaming volume (5 %; water): ≤ 1 ml
Bulk density: about 85
Legal Information
EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 2
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service