Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

07-045

Sigma-Aldrich

Anti-HDAC5 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HDAC5(10014)

Specificity

HDAC5

Immunogen

KLH-conjugated synthetic peptides corresponding to amino acids 536-545 and 194-206 (C-TKTGELPRQP and C-KSKEPTPGGLNHS) of human HDAC5

Application

Detect HDAC5 also known as histone deacetylase 5 with Anti-HDAC5 Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Weaker staining noticed in mouse and rat versus human.

Quality

routinely evaluated by immunoblot on HeLa nuclear extract or HeLa whole cell lysate

Target description

160 kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide.
Format: Purified
Protein A chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

07-045:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteomic analyses of intermediate filaments reveals cytokeratin8 is highly acetylated--implications for colorectal epithelial homeostasis.
Sian H Leech,Caroline A Evans,Leila Shaw,Chi H Wong,Joanne Connolly,John R Griffiths et al.
Proteomics null
Hongbo Ling et al.
Cell cycle (Georgetown, Tex.), 11(20), 3779-3791 (2012-10-02)
Centrosome duplication is controlled both negatively and positively by a number of proteins. The activities and stabilities of those regulatory proteins are in many cases controlled by posttranslational modifications. Although acetylation and deacetylation are highly common posttranslational modifications, their roles
Lauren E Kernochan et al.
Human molecular genetics, 14(9), 1171-1182 (2005-03-18)
Increasing survival motor neuron 2 (SMN2) gene expression may be an effective strategy for the treatment of spinal muscular atrophy (SMA). Histone deacetylase (HDAC) inhibitors have been shown to increase SMN transcript and protein levels, but the specific role of
Oliver H Krämer et al.
The EMBO journal, 22(13), 3411-3420 (2003-07-04)
Histone-modifying enzymes play essential roles in physiological and aberrant gene regulation. Since histone deacetylases (HDACs) are promising targets of cancer therapy, it is important to understand the mechanisms of HDAC regulation. Selective modulators of HDAC isoenzymes could serve as efficient
Syed A H Zaidi et al.
Investigative ophthalmology & visual science, 61(11), 17-17 (2020-09-12)
We determined whether δ-opioid receptor agonist (SNC-121) regulates acetylation homeostasis via controlling histone deacetylases (HDACs) activity and expression in optic nerve head (ONH) astrocytes. ONH astrocytes were treated with SNC-121 (1 µM) for 24 hours. The HDAC activity was measured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service