03-242
RIPAb+ PUM1 Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
pumilio (Drosophila) homolog 1, pumilio 1, pumilio homolog 1 (Drosophila)
About This Item
Recommended Products
biological source
rabbit
antibody form
affinity isolated antibody
clone
polyclonal
purified by
affinity chromatography
species reactivity
rhesus macaque, mouse, human, rat
species reactivity (predicted by homology)
primate (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), bovine (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology)
manufacturer/tradename
RIPAb+
Upstate®
technique(s)
RIP: suitable
immunocytochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... PUM1(9698)
General description
Specificity
Immunogen
Application
Hematopoietic Stem Cells
RNA Metabolism & Binding Proteins
Representative lot data.
H9 human stem cell lysate was resolved by electrophoresis, transferred to PVDF membranes and probed with Anti-Pumilio-1 (PUM1) (1:5,000 dilution).
Proteins were visualized using a Donkey Anti-Rabbit IgG conjugated to HRP and chemiluminescence detection system.
Arrow indicates Pumilio-1 (~120 kDa).
Immunocytochemistry Analysis:
Representative lot data.
Confocal fluorescent analysis of mouse and human embryonic stem cells using Anti-Pumilio-1 (PUM1) (Red). Actin filaments have been labeled with Alexa Fluor™ 488 dye - Phalloidin (Green). Nucleus is stained with DAPI (Blue). This antibody positively stains the cytosol.
Packaging
Quality
Representative lot data.
RIP Lysate prepared from HeLa cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using 2.5 µg of either a normal Rabbit IgG or 2.5 µg Anti-PUM1 antibodyand the Magna RIP® RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700). Successful immunoprecipitation of PUM1-associated RNA was verified by qPCR using RIP Primers p27 mRNA, (Figure 1).
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Target description
UniProtKB/Swiss-Prot Q14671 (PUM1_HUMAN) entry describes 2 isoforms produced by alternative splicing: Isoform 1 at ~126 kDa and Isoform 2 at ~124 kDa
Physical form
Anti-PUM1 (Rabbit Polyclonal). One vial containing 25 µg of affinity purified polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl,with 0.05% sodium azide before the addition of 30% glycerol. Store at -20°C.
Concentration: 0.35 mg/mL
Normal Rabbit IgG. One vial containing 125 µg Rabbit IgG in 125 µL storage buffer containing 0.05% sodium azide. Store at -20°C.
RIP Primers, p27 mRNA. One vial containing 75 μL of each primer (5 μM) specific for a region of the human p27 (Kip1/CDKN1B) mRNA. Store at -20°C.
FOR: CTG CAA CCG ACG ATT CTT CTA CT
REV: GGG CGT CTG CTC CAC AGA
Analysis Note
Includes normal rabbit IgG and primers specific for human p27 mRNA.
Other Notes
Legal Information
Storage Class Code
10 - Combustible liquids
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Please refer to KIT Component information
PRTR
Please refer to KIT Component information
FSL
Please refer to KIT Component information
ISHL Indicated Name
Please refer to KIT Component information
ISHL Notified Names
Please refer to KIT Component information
Cartagena Act
Please refer to KIT Component information
JAN Code
キットコンポーネントの情報を参照してください
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service