Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

860566P

Avanti

12:0 Biotinyl LPA

Avanti Research - A Croda Brand 860566P, powder

Synonym(s):

1-[12-biotinyl(aminododecanoyl)]-2-hydroxy-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H49N4O9PS
CAS Number:
Molecular Weight:
612.72
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860566P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860566P

lipid type

omega FAs

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(O)=O)([H])COC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O.[NH4+]

InChI

1S/C25H46N3O9PS.H3N/c29-19(17-37-38(33,34)35)16-36-23(31)14-8-6-4-2-1-3-5-7-11-15-26-22(30)13-10-9-12-21-24-20(18-39-21)27-25(32)28-24;/h19-21,24,29H,1-18H2,(H,26,30)(H2,27,28,32)(H2,33,34,35);1H3/t19-,20+,21+,24+;/m1./s1

InChI key

ZLKIPNXNKSPNAR-RSYVSERUSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (860566P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

860566P-VAR:
860566P-BULK:
860566P-1MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service