CUP-513
Unsaturated Phosphatidylcholine Liposomes
100% 16:0-18:2 PC
About This Item
Quality Level
composition
Phosphate buffered saline
concentration
10 mM
impurities
100 mol % 16:0-18:2 (Cis) PC (1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphocholine)
particle size
100 nm
pH
7.4
Looking for similar products? Visit Product Comparison Guide
General description
Application
Lipid-protein interactions
Storage and Stability
Liposomes are made under sterile conditions. If you need to take multiple aliquots out of the vial, it is advised to take extreme care in not contaminating the vial. It is recommended to handle the vial under a sterile hood to maintain the sterility of the product. Liposomes should never be frozen. Ice crystals that form during freezing will rupture the lipid membrane of the liposomes and change the size of liposomes particles.
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
CUP-513-2ML:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service