Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

901615

Sigma-Aldrich

Molybdenum sulfide

Single crystalline monolayer flakes on sapphire

Synonym(s):

MoS2 Flakes, Moly sulfide, Monolayer moly sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoS2
UNSPSC Code:
26111700
NACRES:
NA.23

form

film

Looking for similar products? Visit Product Comparison Guide

Application

Owing to the thickness dependent physical and chemical properties, the two dimensional layered inorganic nanomaterials have drawn significant attention recently. They find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc. Transition metal dichalcogenides represent an important class of inorganic 2D materials having aforementioned applications. The CVD grown monolayer films of MoS2, MoSe2, WS2 and WSe2 on insulating substrates such as SiO2 and sapphire are well suited for electronic and photovoltaic devices.

Caution

The material is air and moisture sensitive. Store under inert and moisture free condition.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

901615-1EA:
901615-BULK:
901615-VAR:
901615-1EA-PW:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar J L, et al.
Physical Chemistry, 118, 3847-3847 (2014)
Jianping Shi et al.
ACS nano, 8(10), 10196-10204 (2014-09-12)
Controllable synthesis of monolayer MoS2 is essential for fulfilling the application potentials of MoS2 in optoelectronics and valleytronics, etc. Herein, we report the scalable growth of high quality, domain size tunable (edge length from ∼ 200 nm to 50 μm)
Nikhita D Mansukhani et al.
Small (Weinheim an der Bergstrasse, Germany), 12(3), 294-300 (2015-12-01)
Conditions for the dispersion of molybdenum disulfide (MoS2) in aqueous solution at concentrations up to 0.12 mg mL(-1) using a range of nonionic, biocompatible block copolymers (i.e., Pluronics and Tetronics) are identified. Furthermore, the optimal Pluronic dispersant for MoS2 is
Sheneve Z Butler et al.
ACS nano, 7(4), 2898-2926 (2013-03-08)
Graphene's success has shown that it is possible to create stable, single and few-atom-thick layers of van der Waals materials, and also that these materials can exhibit fascinating and technologically useful properties. Here we review the state-of-the-art of 2D materials
The super materials that could trump graphene.
Elizabeth Gibney
Nature, 522(7556), 274-276 (2015-06-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service