Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

709522

Sigma-Aldrich

3-Quinolineboronic acid

Synonym(s):

(3-Quinolinyl)boronic acid, (3-Quinolyl)boronic acid, Quinolin-3-ylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8BNO2
CAS Number:
Molecular Weight:
172.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

191-196 °C

storage temp.

2-8°C

SMILES string

OB(O)c1cnc2ccccc2c1

InChI

1S/C9H8BNO2/c12-10(13)8-5-7-3-1-2-4-9(7)11-6-8/h1-6,12-13H

InChI key

YGDICLRMNDWZAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain varying amounts of anhydride

Application

Reactant for:
  • Preparation of P1-substituted symmetry-based human immunodeficiency virus protease inhibitors with antiviral activity against drug-resistant viruses
  • Preparation of heterobiaryls via Suzuki-Miyaura cross-coupling with heteroaryl halides in continuous flow
  • Preparation of Ph and heterocyclic(pyridinyl)pyrazolyl)pyridines as TGF-1 and activin A signalling inhibitors
  • Copper-mediated trifluoromethylation
  • Synthesis and bioactivity of combretastatin analogs via regioselective Suzuki coupling of dihaloheteroaromatic compounds
  • Suzuki-Miyaura cross-coupling reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

709522-VAR:
709522-1G:
709522-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manabu Nakazono et al.
Clinica chimica acta; international journal of clinical chemistry, 436, 27-34 (2014-05-13)
Various styrylbenzene compounds were synthesized and evaluated as mainly Aβ amyloid sensors. These compounds, however, cannot be used for detecting amyloid deposition in peripheral nerves because of the inherent sensitivity of the compounds. These compounds often generate false positives especially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service