Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

182907

Sigma-Aldrich

Poly(styrene-co-butadiene)

styrene 45 wt. %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x(CH2CH=CHCH2)y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

slab/chunk

Quality Level

composition

styrene, 45 wt. %

SMILES string

C=CC=C.C=Cc1ccccc1

InChI

1S/C8H8.C4H6/c1-2-8-6-4-3-5-7-8;1-3-4-2/h2-7H,1H2;3-4H,1-2H2

InChI key

MTAZNLWOLGHBHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Molded or extruded into toys, packaging, appliances, communication cassettes, dinnerware and serviceware.

Features and Benefits

Better impact resistance than polystyrene. Amenable to various processing methods. Susceptible to environmental (UV) degradation.

Physical form

High-impact amorphous thermoplastic.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

182907-BULK:
182907-100G:
182907-100G-PW:
182907-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service