Documenti fondamentali
58920-U
Supelco
Rheodyne® Model 7000 Stream Switching Valve
Sinonimo/i:
Rheodyne 7000 Valve
About This Item
Codice UNSPSC:
41115854
eCl@ss:
42079206
NACRES:
SB.51
Caratteristiche
number of ports 6
positions 2
Livello qualitativo
Categorie correlate
Descrizione generale
Designed specifically for column selection, sample clean-up and enrichment, column programming, backflushing, and other stream switching operations. Six peripheral ports (no center port) are interconnected through a two-position rotor. Low volume flow passages (0.6 mm I.D.) minimize dead volume that can affect column switching analyses. The pressure limit is factory set at 5000 psi (350 kg/cm2), but can be adjusted to 7000 psi (490 kg/cm2).
The flat-face seal is easy to adjust.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)