Documenti fondamentali
50647-U
Supelco
Ascentis® Express 90 Å PCS-C18, 2.7 µm HPLC Column
column L × I.D. 100 mm × 4.6 mm, pk of 1 ea
About This Item
Codice UNSPSC:
41115709
NACRES:
SB.52
Confezionamento
pk of 1 ea
Livello qualitativo
Lungh. × diam. int. colonna
100 mm × 4.6 mm
Descrizione generale
Ascentis® Express PCS-C18 columns are specifically engineered to provide efficient separation of basic, acidic, or neutral compounds under low ionic strength (formic acid) mobile phase conditions. These columns contain advanced Fused-Core® particles with pores strategically sized to 90 Å for small molecule analyses. This attribute makes these columns an excellent choice for pharmaceutical method development. The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic compounds as compared to traditional C18 chemistries. This column also provides orthogonal selectivity to other small molecule chemistries.
Risultati analitici
The target analytes include basic compounds.
Note legali
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)