Documenti fondamentali
502561
Upchurch Microtight® Tubing Sleeves
I.D. 0.007 in. (178 μm), yellow
About This Item
Codice UNSPSC:
41115700
Materiali
yellow
Confezionamento
pkg of 10 ea
Diam. int.
0.007 in. (178 μm)
Descrizione generale
Use these PEEK sleeves with all Microtight fittings, to connect capillary tubing. All are 0.025 in. O.D.; normally, choose an I.D. that is 0.002-0.003 in. larger than the O.D. of the capillary tubing. Color-coded.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Microtight is a registered trademark of IDEX Health & Science, LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)