Documenti fondamentali
500410
Supelco
Stand for SCOTTY® 48 Cylinder
About This Item
Codice UNSPSC:
12000000
Confezionamento
pkg of 1 ea
applicazioni
environmental
Descrizione generale
Stabilizes your cylinder on a benchtop or other flat surface. This item will hold cylinders with a four-inch diameter.
Applicazioni
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Note legali
SCOTTY is a registered trademark of Air Liquide
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)