Documenti fondamentali
28523-U
Supelco
ATIS Extraction Glassware with Micro Connector
About This Item
Codice UNSPSC:
41104008
NACRES:
NB.24
Raccordi
connector (20/20)
applicazioni
air monitoring
environmental
industrial hygiene
Descrizione generale
Use the ATIS to thermally extract samples onto an adsorbent tube. The opening in the glassware will accepts solid samples up to 1/2 in. (13mm) in diameter and up to 3" long (76mm) to be inserted into the glassware. The extraction glassware simply slides into the ATIS heating block. Two types of glass joints are available.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)