Documenti fondamentali
28364-U
Supelco
domnick hunter® Air/Nitrogen Generator
Model 3001, 230 volts, output flow, 0 ‑ 3000 cc/min
Sinonimo/i:
NITROX MODEL 3001 N2/AIR GENERATOR WITH COMPRESSOR 220V
About This Item
Codice UNSPSC:
41112500
NACRES:
SB.54
Caratteristiche
output flow 0 ‑ 3000 cc/min
Produttore/marchio commerciale
Domnick Hunter G7-010E
Descrizione generale
Nitrox Air/Nitrogen Generators are ideal for a crowded laboratory! They provide a continuous supply of clean, dry, high-purity nitrogen and clean, high-purity air.
These generators employ pressure swing adsorption technology to produce a continuous supply of high-purity gas. An oil-free air compressor delivers filtered compressed air (7 bar/1015 psig) to a bed of carbon molecular sieve, which selectively removes oxygen, water vapor, and other contaminants. The adsorbent bed alternates between purification and regeneration modes, ensuring product nitrogen meets specified purity levels. A storage reservoir ensures a continuous supply of nitrogen. Additionally, the compressed air is delivered to a bed of activated alumina, to remove water vapor and other contaminants. The adsorbent bed alternates between purification and regeneration modes, and a storage reservoir ensures a continuous supply of pure gas.
With integral Oil-Free Air Compressor
CE approved
Maintenance filter kits and warranty infomation available upon request. Contact Supelco Technical Service at techservice@sial.com or phone 800-359-3041.
Note legali
NITROX is a trademark of domnick hunter group PLC
domnick hunter is a registered trademark of Parker Hannifin
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Contatta l'Assistenza Tecnica