Documenti fondamentali
22161
Supelco
Gas Sampling Bulb
size 125 mL, PTFE stopcock
About This Item
Codice UNSPSC:
41104008
Materiali
PTFE stopcock
tecniche
active air sampling: suitable
Misura
125 mL
applicazioni
air monitoring
environmental
industrial hygiene
Descrizione generale
Gas Sampling Bulb to trap and transport a gas sample, then remove an aliquot (through the plug-type septum) for analysis.
Use these glass bulbs to trap and transport a gas sample, then remove an aliquot (through the plug-type septum) for analysis. Oven annealed to resist damage during transportation and on-site use. Available with PTFE or high vacuum glass stopcocks. One half-hole cylindrical Thermogreen septum is included with each bulb.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)